tert-Butyl (2R,3R)-3-amino-2-methylazetidine-1-carboxylate ≥97%

Supplier: Ambeed
Danger

A693001-1G A693001-250MG A693001-5G
77783-666EA 748 USD
77783-666 77783-667 77783-668
tert-Butyl (2R,3R)-3-amino-2-methylazetidine-1-carboxylate ≥97%
tert-Butyl (2R,3R)-3-amino-2-methylazetidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₂
MDL Number: MFCD30802678
CAS Number: 1932545-45-3
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR