(4-(1H-pyrazol-3-yl)phenyl)boronic acid ≥98%

Supplier: Ambeed
Warning

A692910-100MG A692910-250MG
77783-664EA 169 USD
77783-664 77783-665
(4-(1H-pyrazol-3-yl)phenyl)boronic acid ≥98%
(4-(1H-pyrazol-3-yl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₉BN₂O₂
MDL Number: MFCD15204345
CAS Number: 1226849-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR