Benzyl (2-(2-(2-aminoethoxy)ethoxy)ethyl)carbamate ≥95%

Supplier: Ambeed
Warning

A692674-5G A692674-1G A692674-250MG A692674-25G
77622-464EA 770.44 USD
77622-464 77783-661 77783-662 77783-663
Benzyl (2-(2-(2-aminoethoxy)ethoxy)ethyl)carbamate ≥95%
Benzyl (2-(2-(2-aminoethoxy)ethoxy)ethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₄H₂₂N₂O₄
MW: 282.34 g/mol
MDL Number: MFCD28400772
CAS Number: 169744-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR