(±)-2-Bromo-1-propanol ≥97%

Supplier: Ambeed
Danger

A692395-1G A692395-100MG A692395-250MG
77573-240EA 55 USD
77573-240 77783-659 77783-660
(±)-2-Bromo-1-propanol ≥97%
(±)-2-Bromo-1-propanol

Precaución: for Research Use Only

Formula: HOCH₂BrCHCH₃
MDL Number: MFCD00078663
CAS Number: 598-18-5
UN: 1987
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR