Methyl benzo[b]thiophene-5-carboxylate ≥98%

Supplier: Ambeed
Warning

A692168-1G A692168-5G
77747-390EA 147 USD
77747-390 77747-391
Methyl benzo[b]thiophene-5-carboxylate ≥98%
Methyl benzo[b]thiophene-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈O₂S
MW: 192.23 g/mol
CAS Number: 20532-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR