2-Chloro-3,6-difluorobenzonitrile ≥95%

Supplier: Ambeed
Danger

A691626-25G A691626-1G A691626-5G
77686-674EA 249 USD
77686-674 77901-343 77902-025
2-Chloro-3,6-difluorobenzonitrile ≥95%
2-Chloro-3,6-difluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂ClF₂N
MDL Number: MFCD06660175
CAS Number: 886501-33-3
UN: 3276
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR