(Naphthalen-1-ylmethyl)hydrazine hydrochloride ≥98%

Supplier: Ambeed
Warning

A691168-100MG A691168-1G A691168-250MG
77783-656EA 39 USD
77783-656 77783-657 77783-658
(Naphthalen-1-ylmethyl)hydrazine hydrochloride ≥98%
(Naphthalen-1-ylmethyl)hydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClN₂
MDL Number: MFCD16660416
CAS Number: 94714-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR