D-δ-Tocopherol ≥95%

Supplier: Ambeed
Warning

Synonyms: (2R)-2,8-dimethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol, (+)-δ-Tocopherol, (R)-2,8-Dimethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-ol, Delta-Tocopherol

A690865-1G A690865-5G
77627-400EA 267.8 USD
77627-400 77627-402
D-δ-Tocopherol ≥95%
D-delta-Tocopherol

Precaución: for Research Use Only

Formula: C₂₇H₄₆O₂
MW: 402.663 g/mol
MDL Number: MFCD20486794
CAS Number: 119-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR