3-Amino-5-chloro-N-cyclopropyl-4-methyl-6-(2-(4-methylpiperazin-1-yl)-2-oxoethoxy)thieno[2,3-b]pyridine-2-carboxamide 95+%

Supplier: Ambeed
Warning

Synonyms: LY2119620

A690804-10MG A690804-1MG A690804-5MG A690804-50MG
77614-136EA 59.74 USD
77614-136 77614-138 77614-140 77686-673
3-Amino-5-chloro-N-cyclopropyl-4-methyl-6-(2-(4-methylpiperazin-1-yl)-2-oxoethoxy)thieno[2,3-b]pyridine-2-carboxamide 95+%
3-Amino-5-chloro-N-cyclopropyl-4-methyl-6-(2-(4-methylpiperazin-1-yl)-2-oxoethoxy)thieno[2,3-b]pyridine-2-carboxamide

Precaución: for Research Use Only

Formula: C₁₉H₂₄ClN₅O₃S
MW: 437.94 g/mol
MDL Number: MFCD27996043
CAS Number: 886047-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR