16-Bromo-1-phenyl-2,5,8,11,14-pentaoxahexadecane ≥95%

Supplier: Ambeed
Warning

A690025-1G A690025-250MG A690025-5G
77622-458EA 198.79 USD
77622-458 77622-460 77622-462
16-Bromo-1-phenyl-2,5,8,11,14-pentaoxahexadecane ≥95%
16-Bromo-1-phenyl-2,5,8,11,14-pentaoxahexadecane

Precaución: for Research Use Only

Formula: C₁₇H₂₇BrO₅
MDL Number: MFCD31614133
CAS Number: 675606-48-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR