2-(4-Formylphenyl)propanoic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: Ibuprofen Impurity K

A689820-10MG A689820-1MG A689820-5MG
77617-858EA 28.84 USD
77617-858 77617-860 77617-862
2-(4-Formylphenyl)propanoic acid ≥97%
2-(4-Formylphenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
MW: 178.18 g/mol
MDL Number: MFCD08275588
CAS Number: 43153-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR