1-Boc-6-cyanoindole-2-boronic acid ≥98%

Supplier: Ambeed
Warning

A689653-1G A689653-5G
77873-088EA 50 USD
77873-088 77873-089
1-Boc-6-cyanoindole-2-boronic acid ≥98%
1-Boc-6-cyanoindole-2-boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₅BN₂O₄
MDL Number: MFCD08436056
CAS Number: 913835-67-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR