3-Bromo-6-fluoro-2-methoxypyridine ≥95%

Supplier: Ambeed
Warning

A689629-1G A689629-250MG
77886-609EA 650 USD
77886-609 77886-610
3-Bromo-6-fluoro-2-methoxypyridine ≥95%
3-Bromo-6-fluoro-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrFNO
MDL Number: MFCD18257630
CAS Number: 1256789-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR