3-(Trifluoromethyl)azetidine hydrochloride ≥96%

Supplier: Ambeed
Warning

A689187-250MG A689187-100MG A689187-1G
77573-230EA 87 USD
77573-230 77886-606 77886-607
3-(Trifluoromethyl)azetidine hydrochloride ≥96%
3-(Trifluoromethyl)azetidine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₇ClF₃N
MDL Number: MFCD16140206
CAS Number: 1221272-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR