(2R,3R,4S)-3-Acetamido-2-((1R,2R)-2,3-dihydroxy-1-methoxypropyl)-4-guanidino-3,4-dihydro-2H-pyran-6-carboxylic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: Laninamivir

A688887-100MG A688887-50MG
77900-998EA 327 USD
77900-998 77900-947
(2R,3R,4S)-3-Acetamido-2-((1R,2R)-2,3-dihydroxy-1-methoxypropyl)-4-guanidino-3,4-dihydro-2H-pyran-6-carboxylic acid ≥98%
(2R,3R,4S)-3-Acetamido-2-((1R,2R)-2,3-dihydroxy-1-methoxypropyl)-4-guanidino-3,4-dihydro-2H-pyran-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₂N₄O₇
MDL Number: MFCD18071611
CAS Number: 203120-17-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR