1,2-Difluoro-4,5-dinitrobenzene ≥98%

Supplier: Ambeed
Danger

A688692-10G A688692-1G A688692-25G A688692-5G
77893-378EA 29 USD
77893-378 77893-379 77893-380 77893-381
1,2-Difluoro-4,5-dinitrobenzene ≥98%
1,2-Difluoro-4,5-dinitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂F₂N₂O₄
MDL Number: MFCD00185597
CAS Number: 85686-97-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR