3-Hydroxy-5-(methoxycarbonyl)phenylboronic acid pinacol ester ≥97%

Supplier: Ambeed
Warning

A687594-250MG A687594-5G
77614-124EA 106.09 USD
77614-124 77886-605
3-Hydroxy-5-(methoxycarbonyl)phenylboronic acid pinacol ester ≥97%
3-Hydroxy-5-(methoxycarbonyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₉BO₅
MDL Number: MFCD12546597
CAS Number: 1004294-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR