(S)-2-Amino-3-(4-(4-hydroxy-3-iodophenoxy)-3-iodophenyl)propanoic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: 3,3'-Diiodo-L-thyronine

A687592-1G A687592-250MG
77595-052EA 366.95 USD
77595-052 77673-886
(S)-2-Amino-3-(4-(4-hydroxy-3-iodophenoxy)-3-iodophenyl)propanoic acid ≥95%
(S)-2-Amino-3-(4-(4-hydroxy-3-iodophenoxy)-3-iodophenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₃I₂NO₄
MW: 525.07 g/mol
MDL Number: MFCD01863378
CAS Number: 4604-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR