Muramic acid ≥98%

Supplier: Ambeed
Warning

A687562-10MG A687562-5MG
77673-882EA 619 USD
77673-882 77673-884
Muramic acid ≥98%
Muramic acid

Precaución: for Research Use Only

Formula: C₉H₁₇NO₇
MDL Number: MFCD00213428
CAS Number: 1114-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR