Etidronate disodium ≥98%

Supplier: Ambeed
Warning

A686794-25G A686794-5G A686794-100G A686794-1G
77602-744EA 59.45 USD
77602-744 77602-746 77673-880 77882-545
Etidronate disodium ≥98%
Etidronate disodium

Precaución: for Research Use Only

Formula: C₂H₆Na₂O₇P₂
MDL Number: MFCD00152567
CAS Number: 7414-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR