tert-Butyl (4-chlorobutyl)carbamate ≥98%

Supplier: Ambeed
Warning

A686757-1G A686757-250MG A686757-5G
77627-392EA 137.8 USD
77627-392 77878-522 77878-523
tert-Butyl (4-chlorobutyl)carbamate ≥98%
tert-Butyl (4-chlorobutyl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₈ClNO₂
MDL Number: MFCD11656629
CAS Number: 95388-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR