WYE-125132 ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-(4-(8-Oxa-3-azabicyclo[3.2.1]octan-3-yl)-1-(1,4-dioxaspiro[4.5]decan-8-yl)-1H-pyrazolo[3,4-d]pyrimidin-6-yl)phenyl)-3-methylurea, WYE-132

A686362-1MG A686362-10MG A686362-5MG
77614-122EA 37.08 USD
77614-122 77686-666 77686-667
WYE-125132 ≥98%
WYE-125132

Precaución: for Research Use Only

MW: 519.6 g/mol
MDL Number: MFCD22420813
CAS Number: 1144068-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR