(2-(Cyclopentyloxy)phenyl)boronic acid ≥98%

Supplier: Ambeed
Warning

A685923-1G A685923-250MG
77884-584EA 266 USD
77884-584 77884-585
(2-(Cyclopentyloxy)phenyl)boronic acid ≥98%
(2-(Cyclopentyloxy)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅BO₃
MDL Number: MFCD11183156
CAS Number: 1311159-02-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR