3-(6-Fluoropyridin-3-yl)propanoic acid ≥95%

Supplier: Ambeed
Warning

A685905-100MG A685905-1G
77614-120EA 108.15 USD
77614-120 77686-665
3-(6-Fluoropyridin-3-yl)propanoic acid ≥95%
3-(6-Fluoropyridin-3-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
MDL Number: MFCD18803196
CAS Number: 944998-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR