1-Stearoyl-sn-glycero-3-phosphocholine ≥98%

Supplier: Ambeed
Warning

Synonyms: (R)-2-Hydroxy-3-(stearoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate, (R)-2-Hydroxy-3-(stearoyloxy)propyl (2-(trimethylammonio)ethyl) phosphate

A685884-1G A685884-250MG A685884-5G
77684-591EA 598 USD
77684-591 77684-592 77684-593
1-Stearoyl-sn-glycero-3-phosphocholine ≥98%
1-Stearoyl-sn-glycero-3-phosphocholine

Precaución: For research use only.

Formula: C₂₆H₅₄NO₇P
MW: 523.69 g/mol
MDL Number: MFCD00050427
CAS Number: 19420-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR