(R)-3,3'-Bis(3,5-difluorophenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate ≥97%

Supplier: Ambeed
Warning

Synonyms: (R)-3,3'-Bis(3,5-difluorophenyl)-1,1'-bi-2-naphthol cyclic monophosphate, (11BR)-2,6-bis(3,5-difluorophenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

A684496-100MG A684496-250MG A684496-1G
77617-852EA 71.07 USD
77617-852 77617-854 77747-382
(R)-3,3'-Bis(3,5-difluorophenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate ≥97%
(R)-3,3'-Bis(3,5-difluorophenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate

Precaución: for Research Use Only

Formula: C₃₂H₁₇F₄O₄P
MW: 572.442 g/mol
MDL Number: MFCD28139328
CAS Number: 1191451-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR