3-Fluoro-2-formylbenzeneboronic acid pinacol ester ≥97%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-2-formylphenylboronic acid pinacol ester, 2-Fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde, Benzaldehyde, 2-fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-

A684419-1G A684419-250MG
77686-660EA 94 USD
77686-660 77686-661
3-Fluoro-2-formylbenzeneboronic acid pinacol ester ≥97%
3-Fluoro-2-formylbenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BFO₃
MW: 250.07 g/mol
MDL Number: MFCD18383508
CAS Number: 1246633-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR