6-Methoxyisatoic anhydride ≥98%

Supplier: Ambeed
Warning

Synonyms: 5-Methoxy-2H-3,1-benzoxazine-2,4(1H)-dione, 5-Methoxy-1H-benzo[d][1,3]oxazine-2,4-dione

A684330-1G A684330-5G
77595-046EA 48.18 USD
77595-046 77617-850
6-Methoxyisatoic anhydride ≥98%
6-Methoxyisatoic anhydride

Precaución: for Research Use Only

Formula: C₉H₇NO₄
MW: 193.156 g/mol
MDL Number: MFCD11046419
CAS Number: 67765-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR