(2R)-2-Benzylmorpholine ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A684039-100MG A684039-1G
77684-588EA 45 USD
77684-588 77891-348
(2R)-2-Benzylmorpholine ≥95%, Ambeed.Inc
(2R)-2-Benzylmorpholine

Precaución: For research use only.

Formula: C₁₁H₁₅NO
MDL Number: MFCD14635495
CAS Number: 131887-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR