tert-Butyl isobutyrate ≥95%

Supplier: Ambeed
Danger

A683596-1G A683596-5G
77893-371EA 100 USD
77893-371 77893-372
tert-Butyl isobutyrate ≥95%
tert-Butyl isobutyrate

Precaución: for Research Use Only

Formula: C₈H₁₆O₂
MDL Number: MFCD16038472
CAS Number: 16889-72-8
UN: 3272
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR