tert-Butyl ((1-aminocyclohexyl)methyl)carbamate ≥98%

Supplier: Ambeed
Warning

A683386-1G A683386-100MG A683386-250MG
77573-218EA 87 USD
77573-218 77901-435 77901-150
tert-Butyl ((1-aminocyclohexyl)methyl)carbamate ≥98%
tert-Butyl ((1-aminocyclohexyl)methyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₂₄N₂O₂
MDL Number: MFCD21091404
CAS Number: 1352999-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR