Potassium (2-cyanoethyl)trifluoroborate 95+%

Supplier: Ambeed
Warning

A683375-1G A683375-250MG A683375-100MG A683375-5G
77686-657EA 80 USD
77686-657 77686-658 77901-729 77900-910
Potassium (2-cyanoethyl)trifluoroborate 95+%
Potassium (2-cyanoethyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₃H₄BF₃KN
MDL Number: MFCD11052653
CAS Number: 1159919-79-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR