(R)-6-Oxopiperidine-2-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A683074-1G A683074-250MG
77880-419EA 58 USD
77880-419 77880-420
(R)-6-Oxopiperidine-2-carboxylic acid ≥98%
(R)-6-Oxopiperidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₉NO₃
MDL Number: MFCD09025314
CAS Number: 72002-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR