Bis[2-(di-i-propylphosphino)-4-methylphenyl]amine ≥98%

Supplier: Ambeed
Warning

A682746-250MG A682746-1G A682746-100MG
77617-842EA 341.96 USD
77617-842 77747-379 77747-378
Bis[2-(di-i-propylphosphino)-4-methylphenyl]amine ≥98%
Bis[2-(di-i-propylphosphino)-4-methylphenyl]amine

Precaución: for Research Use Only

Formula: C₂₆H₄₁NP₂
MW: 429.55 g/mol
MDL Number: MFCD29037151
CAS Number: 666856-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR