(S)-Methyl 4-methyl-2-(methylamino)pentanoate hydrochloride ≥95%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Me-Leu-OMe·HCl
A681978-1G
A681978-250MG
77614-106EA
69.01
USD
77614-106
77684-585
(S)-Methyl 4-methyl-2-(methylamino)pentanoate hydrochloride ≥95%
(S)-Methyl 4-methyl-2-(methylamino)pentanoate hydrochloride
Precaución: For research use only.
Formula:
C₈H₁₈ClNO₂ MW: 195.68 g/mol |
MDL Number:
MFCD27578409 CAS Number: 3339-45-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...