1-Ethyl-4-fluorobenzene ≥98%

Supplier: Ambeed
Danger

A681840-1G A681840-5G
77573-214EA 19 USD
77573-214 77878-519
1-Ethyl-4-fluorobenzene ≥98%
1-Ethyl-4-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₉F
MDL Number: MFCD00039218
CAS Number: 459-47-2
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR