Methyl-5-methylthiophene-2-carboxylate ≥98%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-methylthiophene-2-carboxylate

A681318-5G A681318-1G A681318-25G
77673-858EA 68 USD
77673-858 77882-532 77882-533
Methyl-5-methylthiophene-2-carboxylate ≥98%
Methyl-5-methylthiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈O₂S
MDL Number: MFCD06203669
CAS Number: 19432-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR