Cyclohexane-1,4-dicarbaldehyde ≥97%

Supplier: Ambeed
Warning

A680442-1G A680442-5G
77573-204EA 146 USD
77573-204 77614-100
Cyclohexane-1,4-dicarbaldehyde ≥97%
Cyclohexane-1,4-dicarbaldehyde

Precaución: for Research Use Only

Formula: C₈H₁₂O₂
MW: 140.18 g/mol
MDL Number: MFCD18968154
CAS Number: 33424-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR