Sodium 4,5-bis(diphenylphosphino)-9,9-dimethyl-9H-xanthene-2,7-disulfonate ≥98%

Supplier: Ambeed
Warning

A679653-250MG A679653-1G
77747-377EA 83 USD
77747-377 77747-376
Sodium 4,5-bis(diphenylphosphino)-9,9-dimethyl-9H-xanthene-2,7-disulfonate ≥98%
Sodium 4,5-bis(diphenylphosphino)-9,9-dimethyl-9H-xanthene-2,7-disulfonate

Precaución: for Research Use Only

Formula: C₃₉H₃₀Na₂O₇P₂S₂
MW: 782.71 g/mol
MDL Number: MFCD30541777
CAS Number: 215792-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR