2-Fluoro-6-hydroxybenzeneboronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-6-hydroxyphenylboronic acid pinacol ester, 3-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

A679408-250MG A679408-5G
77783-642EA 47 USD
77783-642 77783-643
2-Fluoro-6-hydroxybenzeneboronic acid pinacol ester ≥98%
2-Fluoro-6-hydroxybenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₆BFO₃
MW: 238.06 g/mol
MDL Number: MFCD16994234
CAS Number: 1534369-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR