trans-Aconitic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: Trans-Propene-1,2,3-Tricarboxylic Acid, (E)-Prop-1-ene-1,2,3-tricarboxylic acid

A678863-25G A678863-5G
77602-728EA 43.05 USD
77602-728 77602-730
trans-Aconitic acid ≥98%
trans-Aconitic acid

Precaución: for Research Use Only

Formula: C₆H₆O₆
MW: 174.11 g/mol
MDL Number: MFCD00002721
CAS Number: 4023-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR