m-PEG9-amine ≥98%

Supplier: Ambeed
Warning

Synonyms: 2,5,8,11,14,17,20,23,26-Nonaoxaoctacosan-28-amine

A677170-250MG A677170-100MG
77573-188EA 59 USD
77573-188 77686-645
m-PEG9-amine ≥98%
m-PEG9-amine

Precaución: for Research Use Only

Formula: C₁₉H₄₁NO₉
MDL Number: MFCD16619283
CAS Number: 211859-73-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR