N-Boc-N-methyl-(S)-2-allylglycine ≥95%

Supplier: Ambeed
Warning

A676925-5G A676925-1G A676925-250MG
77783-634EA 209 USD
77783-634 77783-632 77783-633
N-Boc-N-methyl-(S)-2-allylglycine ≥95%
N-Boc-N-methyl-(S)-2-allylglycine

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MW: 229.27 g/mol
MDL Number: MFCD17214445
CAS Number: 136092-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR