4,4'-([2,2'-Bipyridine]-5,5'-diyl)dibenzoic acid ≥98%

Supplier: Ambeed
Warning

A676724-100MG A676724-1G A676724-250MG A676724-5G
77617-818EA 40.17 USD
77617-818 77617-820 77617-822 77749-391
4,4'-([2,2'-Bipyridine]-5,5'-diyl)dibenzoic acid ≥98%
4,4'-([2,2'-Bipyridine]-5,5'-diyl)dibenzoic acid

Precaución: for Research Use Only

Formula: C₂₄H₁₆N₂O₄
MDL Number: MFCD32667111
CAS Number: 1373759-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR