2-(Ethyl(4-((4-nitrophenyl)diazenyl)phenyl)amino)ethanol ≥97%

Supplier: Ambeed
Warning

Synonyms: Disperse Red 1

A676454-5G A676454-1G
77602-720EA 83.03 USD
77602-720 77627-374
2-(Ethyl(4-((4-nitrophenyl)diazenyl)phenyl)amino)ethanol ≥97%
2-(Ethyl(4-((4-nitrophenyl)diazenyl)phenyl)amino)ethanol

Precaución: for Research Use Only

Formula: C₁₆H₁₈N₄O₃
MDL Number: MFCD00007312
CAS Number: 2872-52-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR