Vinorelbine tartrate ≥98%

Supplier: Ambeed
Danger

Synonyms: Methyl (3aR,3a1R,4R,5S,5aR,10bR)-4-acetoxy-3a-ethyl-9-((8R)-4-ethyl-8-(methoxycarbonyl)-1,3,6,7,8,9-hexahydro-2,6-methanoazecino[4,3-b]indol-8-yl)-5-hydroxy-8-methoxy-6-methyl-3a,3a1,4,5,5a,6,11,12-octahydro-1H-indolizino[8,1-cd]carbazole-5-carboxylate bis((2R,3R)-2,3-dihydroxysuccinate), Vinorelbine ditartrate

A676353-100MG A676353-10MG A676353-1MG A676353-250MG A676353-50MG
77669-840EA 230 USD
77669-840 77669-842 77669-844 77669-846 77669-848
Vinorelbine tartrate ≥98%
Vinorelbine tartrate

Precaución: for Research Use Only

Formula: C₅₃H₆₆N₄O₂₀
MDL Number: MFCD03613607
CAS Number: 125317-39-7
UN: 1544
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR