S0859 ≥99%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-N-((2'-(N-cyanosulphamoyl)-[1,1'-biphenyl]-4-yl)methyl)-N-(4-methylbenzyl)benzamide

A676229-10MG A676229-1MG A676229-5MG
77614-090EA 105.06 USD
77614-090 77614-092 77614-094
S0859 ≥99%
S0859

Precaución: for Research Use Only

Formula: C₂₉H₂₄ClN₃O₃S
MW: 530.04 g/mol
MDL Number: MFCD23136018
CAS Number: 1019331-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR