Thiazolo[5,4-b]pyridin-2-amine ≥95%

Supplier: Ambeed
Danger

A676140-1G A676140-250MG
77669-836EA 103 USD
77669-836 77669-838
Thiazolo[5,4-b]pyridin-2-amine ≥95%
Thiazolo[5,4-b]pyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₅N₃S
MW: 151.18 g/mol
MDL Number: MFCD00219102
CAS Number: 31784-70-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR