(9H-Fluoren-9-yl)methyl-4-aminophenylcarbamate ≥97%

Supplier: Ambeed
Warning

Synonyms: (9H-Fluoren-9-yl)methyl (4-aminophenyl)carbamate

A676002-100MG A676002-1G A676002-250MG
77884-567EA 19 USD
77884-567 77884-568 77884-569
(9H-Fluoren-9-yl)methyl-4-aminophenylcarbamate ≥97%
(9H-Fluoren-9-yl)methyl-4-aminophenylcarbamate

Precaución: for Research Use Only

Formula: C₂₁H₁₈N₂O₂
MDL Number: MFCD12912141
CAS Number: 205688-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR