1-(Hydroxymethyl)cyclopropane carboxylic acid ≥98%
Supplier: Ambeed
|
Danger
|
A675790-100MG
A675790-250MG
A675790-1G
A675790-5G
77573-186EA
30
USD
77573-186
77614-088
77673-846
77884-566
1-(Hydroxymethyl)cyclopropane carboxylic acid ≥98%
1-(Hydroxymethyl)cyclopropane carboxylic acid
Precaución: for Research Use Only
Formula:
C₅H₈O₃ |
MDL Number:
MFCD17078844 CAS Number: 49640-66-6 UN: 3261 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...